missing translation for 'onlineSavingsMsg'
Learn More

Silver Hexafluorophosphate 98.0+%, TCI America™

Catalog No. S09815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09815G 5 g
S09811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09815G Supplier TCI America Supplier No. S09815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26042-63-7
Molecular Formula AgF6P
Molecular Weight (g/mol) 252.832
MDL Number MFCD00003415
InChI Key SCQBROMTFBBDHF-UHFFFAOYSA-N
Synonym silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
PubChem CID 168464
IUPAC Name silver;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Ag+]

Specifications

Melting Point 102°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 5 g
UN Number 3260
Formula Weight 252.83
Percent Purity ≥98.0% (T)
Grade Reagent
Identification Passes Test
Appearance White to Light Yellow Powder
Chemical Name or Material Silver Hexafluorophosphate
Show More Show Less
TSCA Yes
Recommended Storage Room Temperature, Cool and dark place, Store under inert gas