missing translation for 'onlineSavingsMsg'
Learn More

Silver Hexafluorophosphate 98.0+%, TCI America™

Catalog No. S09811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09811G 1 g
S09815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09811G Supplier TCI America Supplier No. S09811G

Chemical Identifiers

CAS 26042-63-7
Molecular Formula AgF6P
Molecular Weight (g/mol) 252.832
MDL Number MFCD00003415
InChI Key SCQBROMTFBBDHF-UHFFFAOYSA-N
Synonym silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
PubChem CID 168464
IUPAC Name silver;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Ag+]

Specifications

Melting Point 102°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 1 g
UN Number 3260
Formula Weight 252.83
Percent Purity ≥98.0% (T)
Chemical Name or Material Silver Hexafluorophosphate