missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluoroarsenate, 98+%, Thermo Scientific Chemicals

Catalog No. p-7048833
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1160809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1160809 Supplier Thermo Scientific Chemicals Supplier No. 01160809

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12005-82-2
Molecular Formula AgAsF6
Molecular Weight (g/mol) 296.78
MDL Number MFCD00014142
InChI Key UNSKNPXMHZPWPB-UHFFFAOYSA-N
Synonym silver hexafluoroarsenate, silver hexafluoroarsenate v, silver hexafluoroarsoranuide, acmc-1c478, silver hexakis fluoranyl arsenic 1-, silver 1+ hexafluoro-??-arsanuide, silver 1+ ion hexafluoro-??-arsanuide
PubChem CID 11077325
IUPAC Name silver;hexafluoroarsenic(1-)
SMILES F[As-](F)(F)(F)(F)F.[Ag+]

Specifications

Color White
Physical Form Powder
Quantity 10 g
UN Number UN1557
Formula Weight 296.78
Percent Purity ≥98%
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Silver hexafluoroarsenate
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only