missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Hexafluoroantimonate(V) 97.0+%, TCI America™
Supplier: TCI America S046325G
Specifications
| Silver Hexafluoroantimonate(V) | |
| 26042-64-8 | |
| 25 g | |
| MFCD00003401 | |
| silver hexafluoroantimonate, silver hexafluoroantimonate v, silver hexafluorostiboranuide, silver 1+ hexafluorostibanuide, ag.f6sb, silverhexafluoroantimonate, agsbf6, acmc-209tq8, ksc492g5j, silver i hexafluoroantimonate | |
| F[Sb-](F)(F)(F)(F)F.[Ag+] | |
| 343.619 | |
| 343.62 |
| White | |
| Crystalline Powder | |
| AgF6Sb | |
| 1759 | |
| GSXFODUDOAXUBF-UHFFFAOYSA-H | |
| silver;hexafluoroantimony(1-) | |
| 16687962 | |
| ≥97.0% (T) |
Chemical Identifiers
| 26042-64-8 | |
| 343.619 | |
| GSXFODUDOAXUBF-UHFFFAOYSA-H | |
| 16687962 | |
| F[Sb-](F)(F)(F)(F)F.[Ag+] |
| AgF6Sb | |
| MFCD00003401 | |
| silver hexafluoroantimonate, silver hexafluoroantimonate v, silver hexafluorostiboranuide, silver 1+ hexafluorostibanuide, ag.f6sb, silverhexafluoroantimonate, agsbf6, acmc-209tq8, ksc492g5j, silver i hexafluoroantimonate | |
| silver;hexafluoroantimony(1-) |
Safety and Handling
TSCA : Yes