missing translation for 'onlineSavingsMsg'
Learn More

Silver Hexafluoroantimonate(V) 97.0+%, TCI America™

Catalog No. S04635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04635G 5 g
S046325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04635G Supplier TCI America Supplier No. S04635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26042-64-8
Molecular Formula AgF6Sb
Molecular Weight (g/mol) 343.619
MDL Number MFCD00003401
InChI Key GSXFODUDOAXUBF-UHFFFAOYSA-H
Synonym silver hexafluoroantimonate, silver hexafluoroantimonate v, silver hexafluorostiboranuide, silver 1+ hexafluorostibanuide, ag.f6sb, silverhexafluoroantimonate, agsbf6, acmc-209tq8, ksc492g5j, silver i hexafluoroantimonate
PubChem CID 16687962
IUPAC Name silver;hexafluoroantimony(1-)
SMILES F[Sb-](F)(F)(F)(F)F.[Ag+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula AgSbF6
UN Number 1759
Formula Weight 343.62
Percent Purity ≥97.0% (T)
Chemical Name or Material Silver Hexafluoroantimonate(V)