missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluoroantimonate, 99%

Catalog No. AA1153806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1153806 5 g
AA1153803 1 g
AA1153814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1153806 Supplier Thermo Scientific Chemicals Supplier No. 01153806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26042-64-8 | AgF6Sb | 343.619 g/mol

Silver hexafluoroantimonate is used as a catalyst for the preparation of a number of compounds. It acts as an acidic catalyst in epoxide opening reactions and sulfenylation reactions , controlled initiator in polymerization reactions, and as an activator of catalyst in certain organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26042-64-8
Molecular Formula AgF6Sb
Molecular Weight (g/mol) 343.619
MDL Number MFCD00003401
InChI Key GSXFODUDOAXUBF-UHFFFAOYSA-H
Synonym silver hexafluoroantimonate, silver hexafluoroantimonate v, silver hexafluorostiboranuide, silver 1+ hexafluorostibanuide, ag.f6sb, silverhexafluoroantimonate, agsbf6, acmc-209tq8, ksc492g5j, silver i hexafluoroantimonate
PubChem CID 16687962
IUPAC Name silver;hexafluoroantimony(1-)
SMILES F[Sb-](F)(F)(F)(F)F.[Ag+]

Specifications

Color White to Cream
Physical Form Lump/Powder
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 343.41
Percent Purity 99%
Chemical Name or Material Silver hexafluoroantimonate
Hazard Category H302+H332-H314-H335
Hazard Statement GHS H Statement
H314-H302-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 247-429-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only