missing translation for 'onlineSavingsMsg'
Learn More

Silver gauze, 50 mesh woven from 0.0764mm (0.003in) dia wire

CAS: 7440-22-4 | Ag | 107.87 g/mol

Supplier:  Thermo Scientific Chemicals 040935GT

 View more versions of this product

Catalog No. AA40935GT


Only null left
Explore available promotions

Description

Description

Silver gauze is used for filtration. It is also used in electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver wire gauze
150 x 150 mm
Wire Cloth
MFCD00003397
Insoluble in water.
[Ag]
107.87
CHEBI:9141
7440-22-4
50 Mesh
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only