missing translation for 'onlineSavingsMsg'
Learn More

Silver gauze, 20 mesh woven from 0.356mm (0.014in) dia wire

Filtration, electrodes, contacts | CAS: 7440-22-4 | Ag | 107.87 g/mol

$568.42 - $3571.22

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44449FL
View Documents
Thermo Scientific Chemicals
044449FL
75 x 75 mm
Each for $568.42
Only null left
 
AA44449GT
View Documents
Thermo Scientific Chemicals
044449GT
150 x 150 mm
Each for $1,297.49
Only null left
 
AA44449HB
View Documents
Thermo Scientific Chemicals
044449HB
300 x 300 mm
Each for $3,571.22
Only null left
 
Description

Description

Silver gauze is used for filtration. It is also used in electrodes and contacts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]

Specifications

7440-22-4
20 Mesh
Wire Cloth
MFCD00003397
Insoluble in water.
[Ag]
107.87
CHEBI:9141
75 x 75 mm
Characteristic
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
Silver gauze, 20 mesh woven from 0.356mm (0.014 in.) dia. wire
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only