Learn More
Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
Form: Approx. 3.3g/25 x 25mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol
Supplier: Thermo Scientific Chemicals 039181GH
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver | |
| Annealed | |
| Silver-colored foil | |
| 100 x 100 mm | |
| 99.9% (Metals basis) | |
| Ag | |
| argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
| [Ag] | |
| 107.87 | |
| CHEBI:9141 |
| Silver | |
| 7440-22-4 | |
| 10.5g/cm3 | |
| Characteristic | |
| Foil | |
| MFCD00003397 | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| silver | |
| 23954 |
Chemical Identifiers
| 7440-22-4 | |
| 107.87 | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| 23954 | |
| silver |
| Ag | |
| MFCD00003397 | |
| argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
| CHEBI:9141 | |
| [Ag] |
Safety and Handling
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only