missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)

Form: Approx. 3.3g/25 x 25mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol

Supplier:  Thermo Scientific Chemicals 039181FI

 View more versions of this product

Catalog No. AA39181FI


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver
Annealed
Silver-colored foil
Characteristic
Foil
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
Silver
7440-22-4
50 x 50 mm
99.9% (Metals basis)
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only