missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 0.1mm (0.004in) thick, hard, Premion™, 99.998% (metals basis)

Catalog No. p-4824927
Change view
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog No. Quantity
AA12126FI 50 x 50 mm
AA12126GH 100 x 100 mm
AA12126HP 100 x 500 mm
3 options

Catalog No. AA12126FI

Supplier: Thermo Scientific Chemicals 012126FI

Only null left

Form: Approx. 2.6g/50 x 50mm; Temper: as rolled | CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored foil
Quantity 50 x 50 mm
Odor Characteristic
Form Foil
Assay Percent Range 99.998% (Metals basis)
Chemical Name or Material Silver, Hard
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only