missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis)

Catalog No. p-7048223
Change view
Click to view available options
Quantity:
25 x 150 mm
150 x 150 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA00501FS 25 x 150 mm
AA00501GT 150 x 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA00501FS Supplier Thermo Scientific Chemicals Supplier No. 000501FS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 25 x 150 mm
Odor Characteristic
Weight ≈3g/25x150mm
Form Foil
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Silver foil, 0.075mm (0.003 in.) thick, Hard, Temper: as rolled
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only