missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 0.025mm (0.001in) thick, hard, Premion™, 99.998% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol

$101.34 - $676.11

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11498FF
View Documents
Thermo Scientific Chemicals
011498FF
25 x 25 mm
Each for $101.34
Only null left
 
AA11498FI
View Documents
Thermo Scientific Chemicals
011498FI
50 x 50 mm
Each for $252.44
Only null left
 
AA11498GB
View Documents
Thermo Scientific Chemicals
011498GB
50 x 250 mm
Each for $676.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]

Specifications

7440-22-4
Characteristic
Foil
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
99.998%
Silver foil, 0.025mm (0.001 in.) thick, Hard, Temper: as rolled
25 x 25 mm
≈0.16g/25x25mm
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141
Premion
(metals basis)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only