missing translation for 'onlineSavingsMsg'
Learn More

Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis)

Catalog No. p-4824926
Change view
Click to view available options
Quantity:
150 x 150 mm
150 x 600 mm
150 x 1000 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12190GT 150 x 150 mm
AA12190GV 150 x 600 mm
AA121901EX 150 x 1000 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA12190GT Supplier Thermo Scientific Chemicals Supplier No. 012190GT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Approx. 5.9g/150 x 150mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored foil
Quantity 150 x 150 mm
Odor Characteristic
Form Foil
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Silver, Annealed
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only