missing translation for 'onlineSavingsMsg'
Learn More

Silver Diethyldithiocarbamate, For Analysis, ACS, Reag. Ph Eur Grades, EMSURE™, MilliporeSigma™

Catalog No. M1015150005
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1015150005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1015150005 Supplier Technidata Supplier No. 1.01515.0005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Melting Point 178°C to 183°C
Color Yellow
Physical Form Solid
Quantity 5 g
Linear Formula (C2H5)2NCS2Ag
Formula Weight 256.14. (molar mass)
Grade ACS/Reagent Ph. Eur.
Identification Passes Test
Odor Characteristic odor
Packaging Glass bottle
Appearance Yellow solid
Chemical Name or Material Silver diethyldithiocarbamate
Show More Show Less