missing translation for 'onlineSavingsMsg'
Learn More

Silver Diethyldithiocarbamate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390861
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02003923 10 g Wide Mouth Amber Glass Bottle
02003924 25 g Wide Mouth Amber Glass Bottle
2 options

Catalog No. 02003923

Supplier: Avantor H73902

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Quantity 10 g
Formula Weight 256.14
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Silver Diethyldithiocarbamate