Learn More
Silver diethyldithiocarbamate, ACS
Description
Silver diethyldithiocarbamate cluster plays a useful role for the deposition of acanthite (Ag2S) thin films which is employed in photoelectrochemical applications. Silver diethyldithiocarbamate membrane is used to develop ion selective electrodes for nitrate ion. It is useful in the colorimetric determination of arsenic. It acts as a copper chelator, and is utilized to quantitate copper in biological materials by ESR. It also works as an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 172°C to 175°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 10 g |
| Solubility Information | It is soluble in water and pyridine. |
| Formula Weight | 256.14 |
| Grade | ACS |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Appearance | Yellow to pale brown powder |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 216-003-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only