missing translation for 'onlineSavingsMsg'
Learn More

Silver Diethyldithiocarbamate, ACS Grade, LabChem™

Catalog No. LC224258
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC224258 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC224258 Supplier LabChem Supplier No. LC224258

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Melting Point 175°C to 180°C
Color Yellow
CAS Max % 100
Physical Form Powder
Quantity 25 g
Linear Formula C5H10AgNS2
Formula Weight 256.14
Grade ACS
Identification Passes Test
Packaging Amber Glass
Appearance Yellow Powder
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Silver
Chemical Name or Material Silver Diethyldithiocarbamate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 216-003-7
Recommended Storage Refrigerate