missing translation for 'onlineSavingsMsg'
Learn More

Silver Conductive Ink

Catalog No. p-4408767
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4566106 5 g
AA4566114 25 g
AA4566122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4566106 Supplier Thermo Scientific Chemicals Supplier No. 04566106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Silver Conductive Ink
Density 2.5
Quantity 5 g
Form Liquid
MDL Number MFCD01868501

RUO – Research Use Only