missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride, ultra dry, 99.997% (metals basis)

Catalog No. p-7050143
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3571503 1 g
AA3571506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3571503 Supplier Thermo Scientific Chemicals Supplier No. 03571503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-90-6 | AgCl | 143.32 g/mol

It is used in silver plating, electrode, photographic paper, in making antiseptic silver preparations. It is used to make photographic film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1550°C
Melting Point 455°C
Color White
Physical Form Beads
Quantity 1 g
UN Number UN3077
Merck Index 14,8509
Solubility Information Soluble in alkali cyanides,thiosulfates,ammonium carbonates,ammonium chloride,mercuric nitrate,and silver nitrate. Insoluble in water.
Formula Weight 143.32
Percent Purity 99.997%
Odor Odorless
Packaging Ampule
Refractive Index 2.071
Sensitivity Light sensitive
Density 5.56 g/mL
Assay (metals basis)
Chemical Name or Material Silver chloride, Ultra dry
Show More Show Less
Hazard Category H290
Hazard Statement GHS H Statement
H290
May be corrosive to metals.
Precautionary Statement P234-P390
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 232-033-3
RTECSNumber VW3563000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only