missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride, Premion™, 99.997% (metals basis)

Catalog No. p-7048556
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1085706 5 g
AA1085714 25 g
AA1085722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1085706 Supplier Thermo Scientific Chemicals Supplier No. 01085706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-90-6 | AgCl | 143.32 g/mol

Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements. It serves as an antidote for mercury poisoning, and eliminates mercury from body. It is used in glass manufacturing industry. It is useful in the production of bandages, wound healing products and inglaze lustre, personal deodorant products, as well as for long term preservation of drinking water in water tanks; its pharmaceutical composition finds use as an antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1550°C
Melting Point 455°C
Color White
Physical Form Coarse Powder
Quantity 5 g
UN Number UN3077
Merck Index 14,8509
Solubility Information Practically insoluble in water. Soluble in ammonia,solutions of alkali cyanides,thiosulfates and ammonium carbonates. Insoluble in alcohol and dilute acids.
Formula Weight 143.32
Percent Purity 99.997%
Grade Premion
Odor Odorless
Refractive Index 2.071
Sensitivity Light sensitive
Density 5.56 g/mL
Assay (metals basis)
Chemical Name or Material Silver chloride
Show More Show Less
Hazard Category H290
Hazard Statement GHS H Statement
H290
May be corrosive to metals.
Precautionary Statement P234-P390
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 232-033-3
RTECSNumber VW3563000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only