missing translation for 'onlineSavingsMsg'
Learn More

Silver Chloride, Granular, 99%, Spectrum™ Chemical

Catalog No. p-7166270
Change view
Click to view available options
Quantity:
100 g
25 g
5 g
500 g
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
18613126 5 g Amber Glass Bottle
18613124 25 g Amber Glass Bottle
18613123 100 g Amber Glass Bottle
18613125 500 g Amber Glass Bottle
4 options

Catalog No. 18613126

Supplier: Spectrum Chemical Mfg Cor S10555GM

Only null left

Chemical Identifiers

CAS 7783-90-6

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.