missing translation for 'onlineSavingsMsg'
Learn More

Silver Chloride, Granular, 99%, Spectrum™ Chemical

Catalog No. 18613126
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613126 5 g Amber Glass Bottle
18613124 25 g Amber Glass Bottle
18613123 100 g Amber Glass Bottle
18613125 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613126 Supplier Spectrum Chemical Mfg Cor Supplier No. S10555GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-90-6

Specifications

Melting Point 455°C
CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 5 g
Linear Formula AgCl
Formula Weight 143.32 g/mol
Percent Purity 99%
Grade Reagent
Odor Odorless
Packaging Amber Glass Bottle
Appearance White Granules or Powder
Chemical Name or Material Silver Chloride, Granular
Show More Show Less
Recommended Storage Light Sensitive, Keep tightly closed in light-resistant containers

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.