missing translation for 'onlineSavingsMsg'
Learn More

Silver Chloride, 99%+, MilliporeSigma™

Catalog No. p-4939065
Change view
Click to view available options
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
M1192030025 25 g Plastic bottle
M1192030100 100 g Plastic bottle
2 options

Catalog No. M1192030025

Supplier: Technidata 1.19203.0025

Only null left

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Physical Form Powder
Quantity 25 g
Assay Percent Range 99%+
Formula Weight 143.32
Odor Odorless
Packaging Plastic bottle
Appearance White powder
Chemical Name or Material Silver chloride