missing translation for 'onlineSavingsMsg'
Learn More

Silver Chloride, 99%+, MilliporeSigma™

Catalog No. M1192030025
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1192030025 25 g Plastic bottle
M1192030100 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1192030025 Supplier Technidata Supplier No. 1.19203.0025

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Melting Point 455°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%+
Synonym Silver(1+) chloride
Formula Weight 143.32
Odor Odorless
Packaging Plastic bottle
Appearance White powder
Chemical Name or Material Silver chloride