missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Silver casting grain, 1.6-6.35mm (0.063-0.25in), 99.9% (metals basis)

Catalog No. AA4051114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4051114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4051114 Supplier Thermo Scientific™ Supplier No. 4051114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Jewelry, alloys, electrical contacts

Specifications

CAS 7440-22-4
Quantity 25 g
Form Solid
MDL Number MFCD00003397
Percent Purity 99.9%