Learn More
Silver carbonate, 99%, pure
CAS: 534-16-7 | CAg2O3 | 275.75 g/mol
$256.50 - $737.47
Chemical Identifiers
| CAS | 534-16-7 |
|---|---|
| Molecular Formula | CAg2O3 |
| Molecular Weight (g/mol) | 275.75 |
| MDL Number | MFCD00003403 |
| InChI Key | WIKQEUJFZPCFNJ-UHFFFAOYSA-N |
| Synonym | carbonic acid; silver, carbonic acid disilver |
| PubChem CID | 25203940 |
| IUPAC Name | carbonic acid;silver |
| SMILES | C(=O)(O)O.[Ag].[Ag] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC176970250
|
Thermo Scientific Chemicals
176970250 |
25 g | Glass bottle |
Each for $256.50
|
|
||||
|
AC176971000
|
Thermo Scientific Chemicals
176971000 |
100 g | Glass bottle |
Each for $737.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 534-16-7 | |
| 275.75 | |
| WIKQEUJFZPCFNJ-UHFFFAOYSA-N | |
| 25203940 | |
| C(=O)(O)O.[Ag].[Ag] |
| CAg2O3 | |
| MFCD00003403 | |
| carbonic acid; silver, carbonic acid disilver | |
| carbonic acid;silver |
Specifications
| 210.0°C | |
| Green to Green-Yellow | |
| 25 g | |
| CAg2O3 | |
| MFCD00003403 | |
| 15, 8646 | |
| Solubility in water: insoluble. Other solubilities: soluble in ammonium hydroxide and nitric acid, insoluble in alcohol | |
| C(=O)(O)O.[Ag].[Ag] | |
| 275.75 | |
| 275.75 | |
| Pure | |
| Silver carbonate, 99% |
| 534-16-7 | |
| Granular Powder | |
| 99% | |
| Ag2CO3 | |
| 01,1005; 02,363; 04,425; 06,511; 08,411; 12,432 | |
| carbonic acid; silver, carbonic acid disilver | |
| WIKQEUJFZPCFNJ-UHFFFAOYSA-N | |
| carbonic acid;silver | |
| 25203940 | |
| 99% | |
| Glass bottle |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breath
GHS Signal Word: Danger
EINECSNumber : 208-590-3
RTECSNumber : FG0700000
TSCA : TSCA
RUO – Research Use Only