missing translation for 'onlineSavingsMsg'
Learn More

Silver bromide, Premion™, 99.998% (metals basis)

Catalog No. AA1211022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1211022 100 g
AA1211006 5 g
AA1211014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1211022 Supplier Thermo Scientific Chemicals Supplier No. 01211022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7785-23-1 | AgBr | 187.772 g/mol

Silver bromide is used in photographic film. Used in production of radiograph. It also used in infrared applications as windows and lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7785-23-1
Molecular Formula AgBr
Molecular Weight (g/mol) 187.772
MDL Number MFCD00003398
InChI Key ADZWSOLPGZMUMY-UHFFFAOYSA-M
Synonym silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
PubChem CID 66199
IUPAC Name bromosilver
SMILES Br[Ag]

Specifications

Melting Point 432°C
Color Yellow
Physical Form Crystalline
Quantity 100 g
Odor Odorless
Sensitivity Light sensitive
Appearance Pale yellow powder/flake/crystalline/beads
Assay Percent Range 99.998% (metals basis)
Density 6.47 g/cm3 at 20°C
Solubility Information soluble in alkali cyanide solutions. Partially soluble in ammonia. Insoluble in alcohol and acids. Practically insoluble in water.
Formula Weight 187.78
Chemical Name or Material Silver bromide
Show More Show Less
Hazard Category H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Hazard Statement Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 232-076-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only