missing translation for 'onlineSavingsMsg'
Learn More

Silver Bromide, Granular, Purified, 99%, Spectrum™ Chemical

Catalog No. 18613118
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613118 25 g Amber Glass Bottle
18613119 100 g Amber Glass Bottle
18613120 125 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613118 Supplier Spectrum Chemical Mfg Cor Supplier No. S104825GM

Chemical Identifiers

CAS 7785-23-1

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Purified
Packaging Amber Glass Bottle