missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Behenate 95.0+%, TCI America™
$193.66 - $598.07
Chemical Identifiers
| CAS | 2489-05-6 |
|---|---|
| Molecular Formula | C22H43AgO2 |
| Molecular Weight (g/mol) | 447.45 |
| MDL Number | MFCD00059001 |
| InChI Key | AQRYNYUOKMNDDV-UHFFFAOYSA-M |
| Synonym | silver behenate, silver docosanoate, behenic acid silver salt, unii-n3hp26f3pb, n3hp26f3pb, silver hebenate, docosanoic acid, silver 1+ salt 1:1, docosanoic acid silver salt, docosanoic acid, silver salt, docosanoic acid, silver 1+ salt |
| PubChem CID | 164971 |
| IUPAC Name | silver;docosanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCCCC(=O)[O-].[Ag+] |
Chemical Identifiers
| 2489-05-6 | |
| 447.45 | |
| AQRYNYUOKMNDDV-UHFFFAOYSA-M | |
| 164971 | |
| CCCCCCCCCCCCCCCCCCCCCC(=O)[O-].[Ag+] |
| C22H43AgO2 | |
| MFCD00059001 | |
| silver behenate, silver docosanoate, behenic acid silver salt, unii-n3hp26f3pb, n3hp26f3pb, silver hebenate, docosanoic acid, silver 1+ salt 1:1, docosanoic acid silver salt, docosanoic acid, silver salt, docosanoic acid, silver 1+ salt | |
| silver;docosanoate |
Specifications
| 2489-05-6 | |
| C22H43AgO2 | |
| 5 g | |
| AQRYNYUOKMNDDV-UHFFFAOYSA-M | |
| silver;docosanoate | |
| 164971 | |
| ≥95.0% (T) | |
| Silver Behenate |
| White-Yellow | |
| MFCD00059001 | |
| silver behenate, silver docosanoate, behenic acid silver salt, unii-n3hp26f3pb, n3hp26f3pb, silver hebenate, docosanoic acid, silver 1+ salt 1:1, docosanoic acid silver salt, docosanoic acid, silver salt, docosanoic acid, silver 1+ salt | |
| CCCCCCCCCCCCCCCCCCCCCC(=O)[O-].[Ag+] | |
| 447.45 | |
| 447.45 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0614
TSCA : Yes