missing translation for 'onlineSavingsMsg'
Learn More

Silver Behenate 95.0+%, TCI America™

Catalog No. B03985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03985G 5 g
B039825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B03985G Supplier TCI America Supplier No. B03985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2489-05-6
Molecular Formula C22H43AgO2
Molecular Weight (g/mol) 447.45
MDL Number MFCD00059001
InChI Key AQRYNYUOKMNDDV-UHFFFAOYSA-M
Synonym silver behenate, silver docosanoate, behenic acid silver salt, unii-n3hp26f3pb, n3hp26f3pb, silver hebenate, docosanoic acid, silver 1+ salt 1:1, docosanoic acid silver salt, docosanoic acid, silver salt, docosanoic acid, silver 1+ salt
PubChem CID 164971
IUPAC Name silver;docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)[O-].[Ag+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 447.45
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Silver Behenate
EINECSNumber (2)-0614
TSCA Yes