missing translation for 'onlineSavingsMsg'
Learn More

Silver, 99.9%, foil, 25x25 mm, 1.0 mm thick, 1ea = 6.6gr

CAS: 7440-22-4 | Ag | 107.87 g/mol

$625.46 - $625.46

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC317750040
SDS View Documents
Thermo Scientific Chemicals
317750040
4 Ea.
Each for $625.46
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]

Specifications

Silver
7440-22-4
4 Ea.
Foil
Ag
04,164
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
107.87
Silver, 99.9%, Each sheet is 25x25mm, 6.6g
961.0°C
2212.0°C
99.90%
Ag
MFCD00003397
15, 8643
Solubility in water: insoluble in water. Other solubilities: soluble in hno3, hot h2so4 and alkali cyanide solu, insoluble in alkalies
[Ag]
107.87
CHEBI:9141
99.9%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only