Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Supplier:  Thermo Scientific™ 290MU499
 View more versions of this product
Catalog No. 03150327
Only null left
Description
Description
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columns
Repair broken columns
Specifications
Chemical Identifiers
Fused Silica Capillary Column
Specifications
Capillary Column Connector
290 MU499
32 mm
Fused Silica Capillary Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.