Learn More
Thermo Scientific™ SilTite™ Ferrules
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
Supplier:  Thermo Scientific™ 290MF230
 View more versions of this product
Catalog No. 290MF230
Only null left
Description
Description
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
Specifications
Chemical Identifiers
Fused Silica Capillary GC Columns
SilTite Metal
Ferrule
Specifications
SilTite Metal
Ferrule
For 0.32 mm I.D. Capillary; Pk. of 10
Fused Silica Capillary GC Columns
0.32 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.