missing translation for 'onlineSavingsMsg'
Learn More

New Pig Silicone Water Vacuum Barrier

Catalog No. 17100914
Change view
Click to view available options
Dimensions (L x W x H):
6 x 4 x 2.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
17100914 6 x 4 x 2.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17100914 Supplier New Pig Supplier No. WTR052

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Attach this vacuum barrier to your wet/dry vac to contain and capture water at the same time.

  • Attach the vacuum barrier to a wet/dry vac to contain and capture water.
  • Place the barrier on the floor, connect it to the wet/dry vac, and squeegee water toward it.
  • Compatible with most standard wet/dry vacs, a 2.5 in. hose adapter and end cap are included.
  • Direct water toward the 6 ft. barrier instead of a floor drain or pump.
  • Acts as a backstop to prevent water from spreading.
  • Avoids pump priming and reduces risk of pump failure from dry running.
  • 11 bottom ports remove water quickly.
  • Reusable, clean with soap and water.
  • Suitable for routine washdowns and minor flooding in hospitals, schools, prisons, and property management.

Specifications

Color Black
Material Silicone
Type Spill Blocker
For Use With (Application) Wet Vacuum
Packaging Quantity 1 Ea.
Dimensions (L x W x H) 6 x 4 x 2.25 in.