missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ Silicone Tubing

Catalog No. 04777181
Change view
Click to view available options
Color:
Black
Opaque white
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
04777181 Black
04777187 Opaque white
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04777181 Supplier SOMNI Scientific™ Supplier No. TSB2009
Only null left

Silicone tubing provided custom cut per 12-inch increments to your specification. Silicone tubing is commonly used on anesthesia systems as it handles continuous bending/flexing and yet maintains flexibility over time

  • Silicone tubing provided custom cut per 12 inch increments to your specification
  • Silicone tubing is commonly used on anesthesia systems as it handles continuous bending and flexing
  • Durable
  • Maintains flexibility over time
  • 3/16 in. ID

Specifications

Diameter (English) 3/16 in.
For Use With (Application) Research Animal Inhalant Anesthesia
Material Sillicone
Color Black