missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific Silicone Sealing Mats for Deep Well Plates, 96 Round T110-1, T110-4, and T110-4V
Description
- Effectively seals deep well plates
- Easy to open and close
- Prevents the volatilization of a variety of solvents
- Chemical and heat resistant
- All pre-slit models allow the pipette tip to enter the hole without damage to the silicone cover
- Resistant to DMSO and biological buffers
- Free of RNase, DNase, Pyrogen, and DNA
Specifications
Specifications
| Color | Natural |
| Solvent Or Alcohol Resistant | Alcohol Resistant, Solvent Resistant |
| Material | Silicone |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| Pre-slit | Yes |
| Certified RNase/DNase-free | Yes |
| Pierceable | Yes |
| Temperature Range (Metric) | -80°C to +80°C |
| Show More |