missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Silicone Sealing Mats for Deep Well Plates, 384 Square T110-49

Catalog No. 01183305
Change view
Click to view available options
Material:
Silicone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
01183305 Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01183305 Supplier Simport™ Scientific Supplier No. T11069

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Impermeable sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. These silicone mats are chemical-resistant, reusable, and provide a high degree of confidence.

  • Effectively seals deep well plates
  • Easy to open and close
  • Prevents the volatilization of a variety of solvents
  • Chemical and heat resistant
  • All pre-slit models allow the pipette tip to enter the hole without damage to the silicone cover
  • Resistant to DMSO and biological buffers
  • Free of RNase, DNase, Pyrogen, and DNA

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 384
Pre-slit No
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -80°C to +80°C
Well Shape Square
Fit for Purpose Certified Quality
Show More Show Less