missing translation for 'onlineSavingsMsg'
Learn More

Silicone oil, high temperature

CAS: 63148-58-3

$179.97 - $1712.01

Chemical Identifiers

CAS 63148-58-3
Synonym Poly(methylphenylsiloxane)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC174661000
SDS View Documents
Thermo Scientific Chemicals
174661000
100 g Glass Bottle
Each for $179.97
Only null left
 
AC174665000
SDS View Documents
Thermo Scientific Chemicals
174665000
500 g Glass Bottle
Each for $512.56
Only null left
 
AC174660025
SDS View Documents
Thermo Scientific Chemicals
174660025
2.5 kg Glass Bottle
Each for $1,712.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63148-58-3
Poly(methylphenylsiloxane)

Specifications

63148-58-3
316°C
1.4940 to 1.5010
1.05
Solubility in water: insoluble in water.
Silicone oil
1.0500g/mL
Glass Bottle
100 g
Poly(methylphenylsiloxane)
125 mm2/s (25°C)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only