missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Silicone Oil

Numéro de catalogue. 854091L
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
854091L 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 854091L Fournisseur Sigma Aldrich Code fournisseur 854091L
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil is useful for the preparation of oil baths having a temperature in the range of -50C to +200C.Silicone oil has been used for the protection of carbon steel alloy samples in a study. Silicone oil has also been used as a heat reservoir during NMR (Nuclear Magnetic Resonance) analysis.

Spécifications

CAS 63148-62-9
Density 0.967 g/mL at 20°C
Boiling Point >140°C (0.002 mmHg, lit.)
Refractive Index n20/D 1.403; n20/D 1.406
Linear Formula [-Si(CH3)2O-]n
MDL Number MFCD00132673
Quantity 250 mL