missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Silicone oil

Numéro de catalogue. 3783211L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
3783211L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 3783211L Fournisseur Sigma Aldrich Code fournisseur 3783211L
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil is a low viscosity, liquid polymerized, siloxane with organic side chains. Silicone oil exhibits antimicrobial activity against endopthalmitis causing microorganisms under in vitro conditions.Silicone oil has been used as an intraocular liquid in the treatment of retinal detachments.It may also be used as a two phase partitioning bioreactor to treat gas effluents.

Spécifications

Boiling Point >140°C/0.002, mmHg
CAS 63148-62-9
Quantity 1 L
Linear Formula [-Si(CH3)2 O-]n
MDL Number MFCD00132673
Synonym PDMS; Polydimethylsiloxane
Refractive Index n20/D 1.403 (literature)
Density 0.93 g/mL (at 25°C)