missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Silicone oil

Numéro de catalogue. 3176671L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
3176671L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 3176671L Fournisseur Sigma Aldrich Code fournisseur 3176671L
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil is low viscosity liquid polymerized siloxanes with organic side chains.Low viscosity silicone oil has been used to prepare cellulose based electrorheological particles for medical devices. The diffusive behavior of spreading of a silicone oil polymer on solid surfaces was studied. It was also used to investigate Marangoni flow, induced by two liquid droplets on a solid surface, exposed in air. Many diverse applications of silicone oil are as heat exchangers, hydrophobic coating of glass, ceramics, textiles and leathers, component of salves.

Spécifications

CAS 63148-62-9
Density 0.913 g/mL (at 25°C)
Boiling Point >140°C (0.002 mmHg)
Linear Formula [-Si(CH3)2 O-]n
Refractive Index n20/D 1.403 (literature)
MDL Number MFCD00132673
Quantity 1 L
Synonym PDMS