missing translation for 'onlineSavingsMsg'
Learn More

Electron Microscopy Sciences AFM Substrate Type P Silicon Wafer 6"", 150 mm diameter, <100> Orientation Orientation">

Catalog No. NC1637560
Encompass

These silicon wafers can be used for substrate studies or as substrate for AFM and SEM samples by cleaving the wafer. The wafers are polished on one side.
Diameter: 150 mm
Orientation: <100>
Thickness: 600-690 m
Roughness: 2nm
Resistance: 1-30 ohms

Catalog No. NC1637560 Supplier Electron Microscopy Sciences Supplier No. 7189308

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These silicon wafers can be used for substrate studies or as substrate for AFM and SEM samples by cleaving the wafer. The wafers are polished on one side.
Diameter: 150 mm
Orientation: <100>
Thickness: 600-690 m
Roughness: 2nm
Resistance: 1-30 ohms