Learn More
Silicon oxide, catalyst support, high surface area
Form: S.A. 250m2/g | CAS: 7631-86-9 | O2Si | 60.08 g/mol
$83.68 - $771.44
Chemical Identifiers
| CAS | 7631-86-9 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
| PubChem CID | 24261 |
| ChEBI | CHEBI:30563 |
| IUPAC Name | dioxosilane |
| SMILES | O=[Si]=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4474022
|
Thermo Scientific Chemicals
04474022 |
100 g |
Each for $83.68
|
|
|||||
|
AA4474036
|
Thermo Scientific Chemicals
04474036 |
500 g |
Each for $263.03
|
|
|||||
|
AA44740A3
|
Thermo Scientific Chemicals
044740A3 |
2 kg |
Each for $771.44
|
|
|||||
Description
Typical properties: S.A.250m^2/gSilicon oxide is used as a catalyst support material having a high surface area. It acts as a medium to which catalyst is affixed. Several metals such as platinum, rhodium, ruthenium, silver, copper, nickel, iron, manganese, cobalt, tungsten and gold are supported over silicon dioxide and used as a catalyst in synthetic chemistry for various applications. For example, silica-supported gold catalysts is used for carbon monoxide oxidation by a solution-based technique.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7631-86-9 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| 24261 | |
| dioxosilane |
| O2Si | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| CHEBI:30563 | |
| O=[Si]=O |
Specifications
| White | |
| Pellets | |
| O2Si | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| dioxosilane | |
| 24261 | |
| Odorless | |
| Silicon oxide |
| 7631-86-9 | |
| 100 g | |
| MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
| Insoluble in water. | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:30563 | |
| White pellets |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only