missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, powder, 1.0 micron, 99.9%

Catalog No. p-7046966
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1698614 25 g
AAL1698622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1698614 Supplier Thermo Scientific Chemicals Supplier No. L1698614
Only null left

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silicon(IV) oxide is used in a variety of applications such as manufacturing of glass, ceramics and abrasives, and water filtration. It is a common additive in dentifrice oral care applications and the production of foods, where it is used primarily as an anti-caking and free-flow agent in powdered foods, or to absorb water in hygroscopic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon(IV) oxide is used in a variety of applications such as manufacturing of glass, ceramics and abrasives, and water filtration. It is a common additive in dentifrice oral care applications and the production of foods, where it is used primarily as an anti-caking and free-flow agent in powdered foods, or to absorb water in hygroscopic applications.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
pH 6
Physical Form Powder
Quantity 25 g
Merck Index 14,8493
Solubility Information Insoluble in water.
IUPAC Name dioxosilane
Formula Weight 60.09
Percent Purity 99.9%
Odor Odorless
Density 2
Chemical Name or Material Silicon(IV) oxide, 1.0 micron
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only