missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, amorphous fumed, surface treated, S.A. 105-145m2/g

Catalog No. p-7058578
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4278318 50 g
AA4278330 250 g
AA42783A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4278318 Supplier Thermo Scientific Chemicals Supplier No. 04278318
Only null left

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White
Physical Form Powder
Quantity 50 g
Linear Formula SiO2
IUPAC Name silanedione
Formula Weight 60.09
Odor Odorless
Density 2.2 g/mL
Chemical Name or Material Silicon (IV) oxide, Amorphous fumed, surface treated
Hazard Category Warning
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only