missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, Amorphous fumed, S.A. 175 to 225m2/g

For Adhesives, Coatings, Defomers | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Supplier:  Thermo Scientific Chemicals 04274030

 View more versions of this product

Catalog No. AA4274030


Only null left
Explore available promotions

Description

Description

Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 175 to 225m2/g
Specifications

Specifications

2230°C
1710°C
7631-86-9
250 g
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Insoluble in water.
O=[Si]=O
60.08
CHEBI:30563
Odorless
Silicon(IV) oxide
White
Powder
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
60.09
2.2

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H373: May cause damage to organs through prolonged or repeated exposure.

EINECSNumber : 231-545-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only