missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 99.99% (metals basis)

Catalog No. AA3634622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3634622 100 g
AA3634614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3634622 Supplier Thermo Scientific Chemicals Supplier No. 03634622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Silica is a common additive in the production of foods. An estimated 95% of silicon dioxide produced is consumed in the construction industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Physical Form Fused Lump
Quantity 100 g
Odor Odorless
Assay Percent Range 99.99% (metals basis)
Density 2.2 g/mL to 2.6 g/mL
Merck Index 14,8493
Solubility Information Insoluble in water.
Formula Weight 60.09
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Silicon(IV) oxide
Show More Show Less
EINECSNumber 231-545-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only