missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion, Thermo Scientific Chemicals

Catalog No. AA43091A1
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43091A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA43091A1 Supplier Thermo Scientific Chemicals Supplier No. 043091A1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form Particles in Liquid
Quantity 1 kg
Linear Formula SiO2
Solubility Information Miscible in water. Partially miscible in alcohols,ketones. Immiscible in hydrocarbons.
Formula Weight 60.09
Concentration or Composition (by Analyte or Components) 30% in Ethylene glycol
Packaging Plastic bottle
Specific Gravity 1.3
Density 1.3
Chemical Name or Material Silicon(IV) oxide
Hazard Category H302-H373
Precautionary Statement P260-P264b-P270-P301+P312-P314-P330-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only