missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion, Thermo Scientific Chemicals

$138.58 - $138.58

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43093A1
View Documents
Thermo Scientific Chemicals
043093A1
1 kg
Each for $138.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563

Specifications

White
Particles in Liquid
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
Sweet
Silicon (IV) oxide, Colloidal dispersion
7631-86-9
1 kg
SiO2
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.08
CHEBI:30563
30% in Ethylene glycol
1.3 g/mL
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P301+P312-P314-P330-P501c

H302-H373

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only